siRNA Details
| Gene Symbol: Thap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145929.1 | NP_001139401.1 | 2 | 243 | ||
| NM_175152.4 | NP_780361.2 | 2 | 243 | |||
| GenBank mRNA | AK008225.1 | - | 2 | 215 | ||
| AK038184.1 | - | 2 | 217 | |||
| AK050740.1 | - | 2 | 257 | |||
| BC059091.1 | - | 2 | 228 | |||
| BC094395.1 | - | 2 | 224 | 5700290 | ||
| BY013572.1 | - | 2 | 243 | |||

Thap3
Genomic Map