siRNA Details
| Gene Symbol: Rasip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028544.1 | NP_082820.1 | 5, 6 | 1862 | ||
| GenBank mRNA | AK033276.1 | - | 3, 4 | 1150 | ||
| AK156602.1 | - | 5, 6 | 1874 | |||
| BC072584.1 | - | 2, 3 | 816 | |||
| BC127059.1 | - | 3, 4 | 788 | 40109852 | ||
| BC150766.1 | - | 5, 6 | 1862 | 9087677 | ||

Rasip1
Genomic Map