siRNA Details
| Gene Symbol: Rasip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028544.1 | NP_082820.1 | 4 | 885 | ||
| GenBank mRNA | AK033276.1 | - | 2 | 173 | ||
| AK156602.1 | - | 4 | 897 | |||
| BB646451.1 | - | 2 | 494 | |||
| BC150766.1 | - | 4 | 885 | 9087677 | ||

Rasip1
Genomic Map