siRNA Details
| Gene Symbol: Vrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252447.1 | NP_001239376.1 | 8 | 902 | ||
| NM_027260.3 | NP_081536.2 | 8 | 902 | |||
| GenBank mRNA | AF513620.1 | - | 7 | 566 | ||
| AK012664.1 | - | 8 | 594 | |||
| AK154485.1 | - | 8 | 615 | |||
| AK186520.1 | - | 2 | 60 | |||
| BC013520.1 | - | 7 | 576 | 4236402 | ||

Vrk2
Genomic Map