siRNA Details
| Gene Symbol: Vrk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252447.1 | NP_001239376.1 | 5, 6 | 683 | ||
| NM_027260.3 | NP_081536.2 | 5, 6 | 683 | |||
| GenBank mRNA | AF513620.1 | - | 4, 5 | 347 | ||
| AK012664.1 | - | 5, 6 | 375 | |||
| AK053297.1 | - | 5, 6 | 386 | |||
| AK154485.1 | - | 5, 6 | 396 | |||
| BC013520.1 | - | 4, 5 | 357 | 4236402 | ||

Vrk2
Genomic Map