siRNA Details
| Gene Symbol: Scrn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027268.2 | NP_081544.1 | 6 | 897 | ||
| GenBank mRNA | AK012765.1 | - | 6 | 925 | ||
| AK032573.1 | - | 6 | 924 | |||
| AK050481.1 | - | 6 | 915 | |||
| AK082953.1 | - | 6 | 897 | |||
| AK089968.1 | - | 6 | 887 | |||
| AK129084.1 | - | 6 | 884 | |||
| AK145904.1 | - | 6 | 899 | |||
| BC023889.1 | - | 6 | 884 | 5252046 | ||

Scrn1
Genomic Map