siRNA Details
| Gene Symbol: 2810021J22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172403.2 | NP_765991.1 | 5 | 858 | ||
| GenBank mRNA | AK030099.1 | - | 5 | 883 | ||
| AK134479.1 | - | 5 | 883 | |||
| BC058081.1 | - | 5 | 858 | 6830592 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2810021J22Rik| Gene Symbol: 2810021J22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172403.2 | NP_765991.1 | 5 | 858 | ||
| GenBank mRNA | AK030099.1 | - | 5 | 883 | ||
| AK134479.1 | - | 5 | 883 | |||
| BC058081.1 | - | 5 | 858 | 6830592 | ||