siRNA Details
| Gene Symbol: Ctag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027302.2 | NP_081578.1 | 3 | 663 | ||
| GenBank mRNA | AI503560.1 | - | 1 | 192 | ||
| AK006526.1 | - | 3 | 664 | |||
| BC049681.1 | - | 3 | 657 | 6703247 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ctag2| Gene Symbol: Ctag2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027302.2 | NP_081578.1 | 3 | 663 | ||
| GenBank mRNA | AI503560.1 | - | 1 | 192 | ||
| AK006526.1 | - | 3 | 664 | |||
| BC049681.1 | - | 3 | 657 | 6703247 | ||