siRNA Details
| Gene Symbol: Smc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133786.3 | NP_598547.1 | 22 | 3605 | ||
| GenBank mRNA | AJ534940.2 | - | 21 | 3481 | ||
| BC025542.1 | - | 2 | 277 | |||
| BC061481.1 | - | 22 | 3580 | 6512832 | ||
| BC062939.1 | - | 22 | 3605 | 6841276 | ||

Smc4
Genomic Map