siRNA Details
| Gene Symbol: Dpf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001267625.1 | NP_001254554.1 | 5 | 525 | ||
| NM_001267626.1 | NP_001254555.1 | 5 | 525 | |||
| NM_058212.2 | NP_478119.1 | 5 | 522 | |||
| GenBank mRNA | AF362750.1 | - | 5 | 494 | ||
| AK012970.1 | - | 5 | 530 | |||
| AK039011.1 | - | 5 | 506 | |||
| BC048572.1 | - | 5 | 405 | 6705809 | ||
| FJ151218.1 | - | 5 | 497 | |||

Dpf3
Genomic Map