siRNA Details
| Gene Symbol: 2210016F16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027335.1 | NP_081611.1 | 2 | 549 | ||
| GenBank mRNA | AK052253.1 | - | 2 | 551 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2210016F16Rik| Gene Symbol: 2210016F16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027335.1 | NP_081611.1 | 2 | 549 | ||
| GenBank mRNA | AK052253.1 | - | 2 | 551 | ||