siRNA Details
| Gene Symbol: Ogfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081079.1 | NP_001074548.1 | 6 | 761 | ||
| GenBank mRNA | AK008963.1 | - | 3 | 245 | ||
| BC019747.1 | - | 1 | 82 | |||
| BC041673.1 | - | 5 | 2143 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ogfrl1| Gene Symbol: Ogfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081079.1 | NP_001074548.1 | 6 | 761 | ||
| GenBank mRNA | AK008963.1 | - | 3 | 245 | ||
| BC019747.1 | - | 1 | 82 | |||
| BC041673.1 | - | 5 | 2143 | |||