siRNA Details
| Gene Symbol: Ogfrl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081079.1 | NP_001074548.1 | 7 | 1149 | ||
| GenBank mRNA | AK008963.1 | - | 4 | 633 | ||
| AK037156.1 | - | 1 | 36 | |||
| BC019747.1 | - | 2 | 470 | |||
| BC041673.1 | - | 6 | 2531 | |||

Ogfrl1
Genomic Map