siRNA Details
| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144801.2 | NP_659050.2 | 3 | 299 | ||
| GenBank mRNA | AK010209.1 | - | 3 | 372 | ||
| AK035836.1 | - | 3 | 287 | |||
| AK039173.1 | - | 2 | 369 | |||
| AK087636.1 | - | 2 | 504 | |||
| AK161481.1 | - | 2 | 283 | |||
| AK171908.1 | - | 2 | 2250 | |||
| BB666121.1 | - | 3 | 299 | |||
| BC017618.1 | - | 3 | 293 | 4923610 | ||

Tmem143
Genomic Map