siRNA Details
| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144801.2 | NP_659050.2 | 4, 5 | 563 | ||
| GenBank mRNA | AK039173.1 | - | 3, 4 | 633 | ||
| AK087636.1 | - | 3, 4 | 768 | |||
| BC017618.1 | - | 4, 5 | 557 | 4923610 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem143| Gene Symbol: Tmem143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144801.2 | NP_659050.2 | 4, 5 | 563 | ||
| GenBank mRNA | AK039173.1 | - | 3, 4 | 633 | ||
| AK087636.1 | - | 3, 4 | 768 | |||
| BC017618.1 | - | 4, 5 | 557 | 4923610 | ||