siRNA Details
| Gene Symbol: Poc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027354.2 | NP_081630.2 | 10 | 1396 | ||
| GenBank mRNA | AK081217.1 | - | 581 | |||
| AK154163.1 | - | 10 | 1146 | |||
| BC034901.1 | - | 10 | 1056 | 5255069 | ||
| BE981233.1 | - | 2 | 310 | |||

Poc1a
Genomic Map