siRNA Details
| Gene Symbol: Rnf168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027355.2 | NP_081631.2 | 6 | 1984 | ||
| GenBank mRNA | BC046815.1 | - | 6 | 1956 | 5702430 | |
| BQ572953.1 | - | 1 | 29 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf168| Gene Symbol: Rnf168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027355.2 | NP_081631.2 | 6 | 1984 | ||
| GenBank mRNA | BC046815.1 | - | 6 | 1956 | 5702430 | |
| BQ572953.1 | - | 1 | 29 | |||