siRNA Details
| Gene Symbol: Hdac8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027382.3 | NP_081658.1 | 4 | 399 | ||
| GenBank mRNA | AK011332.1 | - | 4 | 400 | ||
| AK034511.1 | - | 4 | 365 | |||
| AK041965.1 | - | 4 | 398 | |||
| AK131998.1 | - | 4 | 612 | |||
| BC061257.1 | - | 4 | 371 | 30251620 | ||

Hdac8
Genomic Map