siRNA Details
| Gene Symbol: Ppp4r1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114131.1 | NP_001107603.1 | 12 | 1670 | ||
| NM_146081.2 | NP_666193.2 | 12 | 1721 | |||
| GenBank mRNA | AK133969.1 | - | 12 | 1592 | ||
| AK167713.1 | - | 11 | 1132 | |||
| BC026489.1 | - | 4 | 694 | 4504305 | ||

Ppp4r1
Genomic Map