siRNA Details
| Gene Symbol: Secisbp2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177608.3 | NP_808276.2 | 3 | 559 | ||
| GenBank mRNA | AK049924.1 | - | 3 | 560 | ||
| AK145135.1 | - | 3 | 444 | |||
| AK169713.1 | - | 3 | 446 | |||
| AK172920.1 | - | 3 | 428 | |||
| BC095939.1 | - | 3 | 426 | |||
| BC141056.1 | - | 3 | 519 | 9055668 | ||

Secisbp2l
Genomic Map