siRNA Details
| Gene Symbol: Fam135b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177819.3 | NP_808487.2 | 20 | 4261 | ||
| GenBank mRNA | AK005816.1 | - | 8 | 1129 | ||
| BC119174.1 | - | 8 | 877 | |||
| BC137579.1 | - | 8 | 877 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam135b| Gene Symbol: Fam135b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177819.3 | NP_808487.2 | 20 | 4261 | ||
| GenBank mRNA | AK005816.1 | - | 8 | 1129 | ||
| BC119174.1 | - | 8 | 877 | |||
| BC137579.1 | - | 8 | 877 | |||