siRNA Details
| Gene Symbol: Gpatch2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027405.2 | NP_081681.2 | 2 | 230 | ||
| GenBank mRNA | AK006185.1 | - | 2 | 231 | ||
| AK031712.1 | - | 2 | 234 | |||
| AK129299.1 | - | 2 | 207 | |||
| AK147916.1 | - | 2 | 219 | |||
| BC048169.1 | - | 2 | 239 | 6821902 | ||
| BC050782.1 | - | 2 | 210 | |||
| BC058256.1 | - | 2 | 230 | 6466603 | ||

Gpatch2l
Genomic Map