siRNA Details
| Gene Symbol: Tecpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027410.1 | NP_081686.1 | 16 | 2535 | ||
| GenBank mRNA | AB093295.1 | - | 15 | 2704 | ||
| AK032858.1 | - | 16 | 2509 | |||
| AK169888.1 | - | 16 | 2482 | |||
| AK170108.1 | - | 16 | 2482 | |||
| AK179982.1 | - | 2 | 33 | |||
| AK184052.1 | - | 1 | 30 | |||
| AK195038.1 | - | 1 | 31 | |||
| AK203696.1 | - | 253 | ||||
| BC025567.1 | - | 3 | 280 | |||
| BC046968.1 | - | 16 | 2535 | 6306206 | ||

Tecpr1
Genomic Map