siRNA Details
| Gene Symbol: Asnsd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133728.3 | NP_598489.2 | 4 | 1799 | ||
| GenBank mRNA | AK031295.1 | - | 4 | 1757 | ||
| AK034746.1 | - | 1718 | ||||
| AK049194.1 | - | 4 | 1746 | |||
| AK050045.1 | - | 4 | 1739 | |||
| AK076793.1 | - | 4 | 1701 | |||
| AK155030.1 | - | 4 | 1738 | |||
| AK168103.1 | - | 4 | 1739 | |||
| AK170510.1 | - | 4 | 1715 | |||
| BC013617.1 | - | 4 | 1619 | 3979452 | ||

Asnsd1
Genomic Map