siRNA Details
| Gene Symbol: Ints2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027421.2 | NP_081697.2 | 21 | 3394 | ||
| GenBank mRNA | AK013101.1 | - | 2 | 376 | ||
| AK129323.1 | - | 21 | 3376 | |||
| BC050081.1 | - | 21 | 3170 | 30023969 | ||
| BC059056.1 | - | 20 | 3246 | 6821329 | ||
| BC118007.1 | - | 21 | 3109 | 40086155 | ||

Ints2
Genomic Map