siRNA Details
| Gene Symbol: Polr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027423.1 | NP_081699.1 | 14, 15 | 1500 | ||
| GenBank mRNA | AK039366.1 | - | 14, 15 | 1578 | ||
| BC068143.1 | - | 14, 15 | 1500 | 30362434 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Polr3b| Gene Symbol: Polr3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027423.1 | NP_081699.1 | 14, 15 | 1500 | ||
| GenBank mRNA | AK039366.1 | - | 14, 15 | 1578 | ||
| BC068143.1 | - | 14, 15 | 1500 | 30362434 | ||