siRNA Details
| Gene Symbol: Rufy2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027425.3 | NP_081701.2 | 11 | 1061 | ||
| GenBank mRNA | AK011847.1 | - | 11 | 1031 | ||
| AK083205.1 | - | 11 | 1046 | |||
| AK173195.1 | - | 11 | 1022 | |||
| BC017645.1 | - | 11 | 1023 | |||
| BC038265.1 | - | 10 | 932 | |||
| BC137736.1 | - | 11 | 1010 | 8860755 | ||
| BC137737.1 | - | 11 | 1010 | 8860756 | ||

Rufy2
Genomic Map