siRNA Details
| Gene Symbol: Inf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198411.2 | NP_940803.2 | 17 | 2744 | ||
| GenBank mRNA | BC024095.1 | - | 10 | 886 | ||
| BC060610.1 | - | 10 | 995 | 6416724 | ||
| BC099931.1 | - | 10 | 892 | 30063439 | ||
| BC115422.1 | - | 10 | 965 | |||
| BC115423.1 | - | 10 | 965 | |||
| DQ834374.1 | - | 17 | 2752 | |||

Inf2
Genomic Map