siRNA Details
| Gene Symbol: Inf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198411.2 | NP_940803.2 | 18 | 2959 | ||
| GenBank mRNA | BC024095.1 | - | 11 | 1101 | ||
| BC048907.1 | - | 1 | 37 | |||
| BC060610.1 | - | 11 | 1210 | 6416724 | ||
| BC099931.1 | - | 11 | 1107 | 30063439 | ||
| BC115422.1 | - | 11 | 1180 | |||
| BC115423.1 | - | 11 | 1180 | |||
| DQ834374.1 | - | 18 | 2967 | |||

Inf2
Genomic Map