siRNA Details
| Gene Symbol: Cd248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_054042.2 | NP_473383.1 | 1 | 2236 | ||
| GenBank mRNA | AF378758.1 | - | 1 | 2194 | ||
| AF388572.1 | - | 1 | 2191 | |||
| AK011844.1 | - | 1 | 1062 | |||
| BC046318.1 | - | 1 | 2236 | 6314798 | ||
| BC128295.1 | - | 1 | 333 | |||
| BC152362.1 | - | 1 | 333 | |||

Cd248
Genomic Map