siRNA Details
| Gene Symbol: Mipep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027436.3 | NP_081712.2 | 11 | 1346 | ||
| GenBank mRNA | AK017491.1 | - | 11 | 1322 | ||
| AK038479.1 | - | 11 | 1340 | |||
| AK044460.1 | - | 11 | 1331 | |||
| AK076613.1 | - | 11 | 1276 | |||
| AK146428.1 | - | 12 | 1337 | |||
| BC049871.1 | - | 2 | 186 | |||
| BC110358.1 | - | 11 | 1324 | 5322591 | ||
| BC144798.1 | - | 11 | 1327 | |||
| BC145711.1 | - | 11 | 1327 | 40131014 | ||

Mipep
Genomic Map