siRNA Details
| Gene Symbol: Atp6ap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027439.4 | NP_081715.1 | 4, 5 | 536 | ||
| GenBank mRNA | AB192471.2 | - | 4, 5 | 412 | ||
| AK017482.1 | - | 4, 5 | 468 | |||
| AK029405.1 | - | 4, 5 | 439 | |||
| AK076495.1 | - | 4, 5 | 481 | |||
| AK151735.1 | - | 4, 5 | 467 | |||
| AK168139.1 | - | 4, 5 | 493 | |||
| AW985491.1 | - | 1, 2 | 103 | |||
| BC014706.1 | - | 4, 5 | 407 | 4457079 | ||

Atp6ap2
Genomic Map