siRNA Details
| Gene Symbol: Qpct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027455.2 | NP_081731.1 | 5 | 1157 | ||
| GenBank mRNA | AK017598.1 | - | 5 | 820 | ||
| AK037197.1 | - | 5 | 810 | |||
| AK045974.1 | - | 5 | 1158 | |||
| AK148394.1 | - | 5 | 828 | |||
| BC020023.1 | - | 4 | 728 | 3491756 | ||
| BC151027.1 | - | 5 | 901 | 9087940 | ||
| BC151029.1 | - | 5 | 901 | 9087942 | ||

Qpct
Genomic Map