siRNA Details
| Gene Symbol: Tmem242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027457.4 | NP_081733.3 | 3 | 322 | ||
| GenBank mRNA | AK003559.2 | - | 3 | 321 | ||
| AK009851.1 | - | 3 | 305 | |||
| AK017626.1 | - | 3 | 319 | |||
| AK043276.1 | - | 3 | 290 | |||
| AK088025.1 | - | 3 | 302 | |||
| AK155326.1 | - | 3 | 301 | |||
| AK158107.1 | - | 3 | 301 | |||
| BC019420.1 | - | 3 | 286 | 5053733 | ||
| BY013779.1 | - | 3 | 323 | |||

Tmem242
Genomic Map