siRNA Details
| Gene Symbol: Tmem242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027457.4 | NP_081733.3 | 3, 4 | 373 | ||
| GenBank mRNA | AK009851.1 | - | 3, 4 | 356 | ||
| AK043276.1 | - | 3, 4 | 341 | |||
| AK158107.1 | - | 3, 4 | 352 | |||
| BC019420.1 | - | 3, 4 | 337 | 5053733 | ||

Tmem242
Genomic Map