siRNA Details
| Gene Symbol: Tmtc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028651.2 | NP_082927.1 | 12, 13 | 1671 | ||
| GenBank mRNA | AK015064.1 | - | 2, 3 | 249 | ||
| AK017580.1 | - | 2, 3 | 200 | |||
| AK028333.1 | - | 12, 13 | 1652 | |||
| AK033947.1 | - | 12, 13 | 1672 | |||
| AK036741.1 | - | 11, 12 | 1559 | |||
| AK043293.1 | - | 12, 13 | 1698 | |||
| AK044391.1 | - | 12, 13 | 1627 | |||
| AK045271.1 | - | 12, 13 | 1633 | |||
| AK049748.1 | - | 13, 14 | 1812 | |||
| AK082526.1 | - | 11, 12 | 1499 | |||
| BC031368.1 | - | 8, 9 | 1045 | 5029461 | ||

Tmtc4
Genomic Map