siRNA Details
| Gene Symbol: Tmtc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_028651.2 | NP_082927.1 | 9 | 1242 | ||
| GenBank mRNA | AK028333.1 | - | 9 | 1223 | ||
| AK033947.1 | - | 9 | 1243 | |||
| AK039093.1 | - | 9 | 1187 | |||
| AK043293.1 | - | 9 | 1269 | |||
| AK044391.1 | - | 9 | 1198 | |||
| AK045271.1 | - | 9 | 1204 | |||
| AK049748.1 | - | 10 | 1383 | |||
| AK082526.1 | - | 8 | 1070 | |||
| BC031368.1 | - | 5 | 616 | 5029461 | ||

Tmtc4
Genomic Map