siRNA Details
| Gene Symbol: Fra10ac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081075.1 | NP_001074544.1 | 3 | 264 | ||
| GenBank mRNA | AK017666.1 | - | 3 | 326 | ||
| AK077572.1 | - | 3 | 237 | |||
| AK172241.1 | - | 3 | 271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fra10ac1| Gene Symbol: Fra10ac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081075.1 | NP_001074544.1 | 3 | 264 | ||
| GenBank mRNA | AK017666.1 | - | 3 | 326 | ||
| AK077572.1 | - | 3 | 237 | |||
| AK172241.1 | - | 3 | 271 | |||