siRNA Details
| Gene Symbol: Gfod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027469.4 | NP_081745.1 | 3 | 1140 | ||
| GenBank mRNA | AK017684.1 | - | 3 | 1141 | ||
| AK141521.1 | - | 1150 | ||||
| BC013623.1 | - | 3 | 1152 | 3969153 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gfod2| Gene Symbol: Gfod2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027469.4 | NP_081745.1 | 3 | 1140 | ||
| GenBank mRNA | AK017684.1 | - | 3 | 1141 | ||
| AK141521.1 | - | 1150 | ||||
| BC013623.1 | - | 3 | 1152 | 3969153 | ||