siRNA Details
| Gene Symbol: Filip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081243.1 | NP_001074712.1 | 5 | 3254 | ||
| GenBank mRNA | BC125581.1 | - | 5 | 3099 | 40130096 | |
| BC131965.1 | - | 5 | 3099 | 40130242 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Filip1| Gene Symbol: Filip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081243.1 | NP_001074712.1 | 5 | 3254 | ||
| GenBank mRNA | BC125581.1 | - | 5 | 3099 | 40130096 | |
| BC131965.1 | - | 5 | 3099 | 40130242 | ||