siRNA Details
| Gene Symbol: Filip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081243.1 | NP_001074712.1 | 2 | 508 | ||
| GenBank mRNA | AK017709.1 | - | 2 | 323 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Filip1| Gene Symbol: Filip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081243.1 | NP_001074712.1 | 2 | 508 | ||
| GenBank mRNA | AK017709.1 | - | 2 | 323 | ||