siRNA Details
| Gene Symbol: Ecd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027475.3 | NP_081751.1 | 6 | 1069 | ||
| GenBank mRNA | AK017681.1 | - | 4 | 493 | ||
| AK028664.1 | - | 6 | 1033 | |||
| AK029169.1 | - | 5 | 2260 | |||
| AK043261.1 | - | 6 | 918 | |||
| BC013470.1 | - | 6 | 1018 | 4220504 | ||

Ecd
Genomic Map