siRNA Details
| Gene Symbol: Mutyh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159581.1 | NP_001153053.1 | 10, 11 | 827 | ||
| NM_133250.2 | NP_573513.2 | 9, 10 | 734 | |||
| GenBank mRNA | AB117937.1 | - | 7, 8 | 622 | ||
| AB117938.1 | - | 8, 9 | 696 | |||
| AK049542.1 | - | 1, 2 | 76 | |||
| AK077546.1 | - | 9, 10 | 731 | |||
| AK156790.1 | - | 10, 11 | 817 | |||
| AY007717.1 | - | 8, 9 | 690 | |||
| BC057942.1 | - | 9, 10 | 715 | 5320744 | ||

Mutyh
Genomic Map