siRNA Details
| Gene Symbol: Fbxo33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033156.4 | NP_001028328.2 | 3 | 1187 | ||
| GenBank mRNA | AK132233.1 | - | 1 | 1146 | ||
| BC006669.1 | - | 1 | 323 | |||
| BC020022.1 | - | 3 | 1156 | |||
| BC025217.1 | - | 3 | 767 | |||
| BC138618.1 | - | 1 | 1121 | 8861638 | ||
| BC138621.1 | - | 1 | 1121 | 8861641 | ||

Fbxo33
Genomic Map