siRNA Details
| Gene Symbol: Tmem144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027495.4 | NP_081771.2 | 2 | 402 | ||
| GenBank mRNA | AK017808.1 | - | 2 | 421 | ||
| AK079384.1 | - | 2 | 403 | |||
| BC018493.1 | - | 2 | 310 | 3593194 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem144| Gene Symbol: Tmem144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027495.4 | NP_081771.2 | 2 | 402 | ||
| GenBank mRNA | AK017808.1 | - | 2 | 421 | ||
| AK079384.1 | - | 2 | 403 | |||
| BC018493.1 | - | 2 | 310 | 3593194 | ||