siRNA Details
| Gene Symbol: Prdm16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177995.1 | NP_001171466.1 | 8 | 1291 | ||
| NM_001291026.1 | NP_001277955.1 | 8 | 1285 | |||
| NM_001291029.1 | NP_001277958.1 | 8 | 1291 | |||
| NM_027504.3 | NP_081780.3 | 8 | 1288 | |||
| GenBank mRNA | AB078338.1 | - | 8 | 1180 | ||
| AK083829.1 | - | 8 | 1175 | |||
| BC053082.1 | - | 8 | 1226 | |||
| BC059838.1 | - | 8 | 1269 | 6409778 | ||

Prdm16
Genomic Map