siRNA Details
| Gene Symbol: Vcpip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK017812.1 | - | 1 | 158 | ||
| AK030104.1 | - | 1 | 1954 | |||
| AK122563.1 | - | 1 | 1967 | |||
| AK144688.1 | - | 1 | 1977 | |||
| BC006059.1 | - | 1 | 1939 | |||
| BC052908.1 | - | 1 | 144 | |||
| BC059209.1 | - | 1 | 1973 | 6406239 | ||

Vcpip1
Genomic Map