siRNA Details
| Gene Symbol: Prdm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027547.2 | NP_081823.2 | 4 | 599 | ||
| GenBank mRNA | AK014501.1 | - | 4 | 523 | ||
| BC138901.1 | - | 4 | 573 | 8861923 | ||
| BC138902.1 | - | 4 | 573 | 8861924 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prdm5| Gene Symbol: Prdm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027547.2 | NP_081823.2 | 4 | 599 | ||
| GenBank mRNA | AK014501.1 | - | 4 | 523 | ||
| BC138901.1 | - | 4 | 573 | 8861923 | ||
| BC138902.1 | - | 4 | 573 | 8861924 | ||