siRNA Details
| Gene Symbol: Prdm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027547.2 | NP_081823.2 | 12 | 1525 | ||
| GenBank mRNA | AK014501.1 | - | 12 | 1449 | ||
| AK018310.1 | - | 8 | 911 | |||
| AK021384.1 | - | 3 | 334 | |||
| BC138901.1 | - | 12 | 1499 | 8861923 | ||
| BC138902.1 | - | 12 | 1499 | 8861924 | ||
| BG070775.2 | - | 4 | 653 | |||
| BX511446.1 | - | 1 | 28 | |||

Prdm5
Genomic Map