siRNA Details
| Gene Symbol: Rasl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033158.2 | NP_001028330.2 | 4 | 360 | ||
| NM_001162923.1 | NP_001156395.1 | 4 | 360 | |||
| GenBank mRNA | AK014511.1 | - | 4 | 323 | ||
| AK048176.1 | - | 4 | 314 | |||
| AK087136.1 | - | 5 | 848 | |||
| AK171623.1 | - | 4 | 316 | |||
| BC125036.1 | - | 4 | 337 | 40102054 | ||

Rasl12
Genomic Map